Hydrazinium monochloride 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hydrazinium monochloride, Hydrazinium chloride, Hydrazine monohydrochloride, Hydrazine hydrochloride

20199-0010 20199-0051 20199-2500
TS20199-0010EA 230 USD
TS20199-0010 TS20199-0051 TS20199-2500
Hydrazinium monochloride 98+%
Hydrazinium monochloride
Formula: H₅ClN₂
Melting Pt: 89…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00044368
CAS Number: 2644-70-4
EINECS: 220-154-4
UN: 3288
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder or crystals
Infrared spectrum Conforms
Titration Argentometric ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR