Pyridinium dichromate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: PDC

20088-0010 20088-0500 20088-2500
TS20088-0010EA 357 USD
TS20088-0010 TS20088-0500 TS20088-2500
Pyridinium dichromate 98%
Pyridinium dichromate
Formula: C₁₀N₁₂Cr₂N₂O₇
MW: 532.19 g/mol
Melting Pt: 152…153 °C
Storage Temperature: Ambient
MDL Number: MFCD00013105
CAS Number: 20039-37-6
EINECS: 243-478-8
UN: 3085
ADR: 5.1 II

Order Now

Specification Test Results

Appearance Orange to brown Crystalline powder
Infrared spectrum Conforms
Titration Iodometric ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR