Carbol fuchsin, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-[(4-Amino-m-tolyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]-o-toluidine hydrochloride

19579-0050 19579-0250 19579-1000
TS19579-0050EA 62 USD
TS19579-0050 TS19579-0250 TS19579-1000
Carbol fuchsin, pure
Carbol fuchsin
Formula: C₂₁H₂₂ClN₃
MW: 351.88 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013129
CAS Number: 4197-24-4
EINECS: 224-086-6
UN: 2923
ADR: 8,III

Order Now

Specification Test Results

Appearance Dark green Crystalline powder
Infrared spectrum Conforms
UV spectrum Complies
UV Lambda max (in water) about 542 nm
Suitability (for microscopy) Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR