Celestine blue, pure

Supplier: Thermo Scientific Chemicals

Synonyms: 7-(diethylylidene)-3,4-dihydroxyphenoxazinecarboxamide, chloride, 1-Carbamoyl-7-(diethylamino)-3,4-dihydroxyphenoxazinium chloride, Phenoxazin-5-ium,1-(aminocarbonyl)-7-(diethylamino)-3,4-dihydroxy-, chloride (1:1)

19556-0100 19556-0500
TS19556-0100EA 100 USD
TS19556-0100 TS19556-0500
Celestine blue, pure
Celestine blue
Formula: C₁₇H₁₈ClN₃O₄
MW: 363.798 g/mol
Melting Pt: 227…230 °C
Storage Temperature: Ambient
MDL Number: MFCD00011927
CAS Number: 1562-90-9

Order Now

Specification Test Results

Appearance Dark grey to dark brown to black Powder
Infrared spectrum Conforms
UV spectrum Complies
UV Lambda max 638 to 648 nm
Specific extinction E (1%/1cm) ≥700 (in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR