Chromotrope 2R, pure

Supplier: Thermo Scientific Chemicals
Warning

19201-0250 19201-1000
TS19201-0250EA 57 USD
TS19201-0250 TS19201-1000
Chromotrope 2R, pure
Chromotrope 2R
Formula: C₁₆H₁₀N₂Na₂O₈S₂
Storage Temperature: Ambient
MDL Number: MFCD00003955
CAS Number: 4197-07-3
EINECS: 224-085-0

Order Now

Specification Test Results

Appearance Red-brown Crystalline powder
Infrared spectrum Conforms
Specific extinction E (1%/1cm) ≥360 (509 nm, water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR