Sudan red 7B (fat red bluish, solvent red 19, fat red 7B)
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Ethyl-1-((4-(phenyldiazenyl)phenyl)diazenyl)naphthalen-2-amine
19163-0050
19163-0500
TS19163-0050EA
36
USD
TS19163-0050
TS19163-0500
Sudan red 7B (fat red bluish, solvent red 19, fat red 7B)
Sudan red 7B (fat red bluish, solvent red 19, fat red 7B)
Formula:
C₂₄H₂₁N₅ MW: 379.4570 g/mol Melting Pt: 130 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00003904 CAS Number: 6368-72-5 EINECS: 228-862-5 |
Specification Test Results
Appearance | Bordeaux Crystalline powder |
Infrared spectrum | Conforms |
UV spectrum | Complies |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...