Carmine alum lake of carminic acid, pure, high purity biological stain

Supplier: Thermo Scientific Chemicals

Synonyms: Carminic acid, Cochineal, Alum lake of Carminic Acid, Natural Red 4, FD and C Carmine, CI 75470, Carmine, calcium/aluminum lake

19020-0050 19020-0250 19020-1000
TS19020-0050EA 50 USD
TS19020-0050 TS19020-0250 TS19020-1000
Carmine alum lake of carminic acid, pure, high purity biological stain
Carmine
Formula: C₂₂H₂₀O₁₃
Storage Temperature: Ambient
MDL Number: MFCD00167028
CAS Number: 1390-65-4
EINECS: 215-724-4

Order Now

Specification Test Results

Appearance Dark red to purple Powder
Infrared spectrum Conforms
UV spectrum Complies
Assay ≥42.0 % (Carminic acid)
Loss on drying ≤20 %
Ash ≤12 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR