Victoria blue B, pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Basic blue 26, N-(4-((4-(Dimethylamino)phenyl)(4-(phenylamino)naphthalen-1-yl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanaminium chloride

19019-0250 19019-1000
TS19019-0250EA 50 USD
TS19019-0250 TS19019-1000
Victoria blue B, pure
Victoria Blue B
Formula: C₃₃H₃₂ClN₃
MW: 506.803 g/mol
Melting Pt: 206 °C
Storage Temperature: Ambient
MDL Number: MFCD00011878
CAS Number: 2580-56-5
EINECS: 219-943-6

Order Now

Specification Test Results

Appearance Dark brown Glistening crystalline powder
Infrared spectrum Conforms
UV spectrum Complies
Loss on drying ≤5 % (60°C)
UV Lambda max 599 to 605 nm (in ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR