Patent Blue VF (Disulfine blue), pure indicator grade
Supplier: Thermo Scientific Chemicals
Synonyms:
acid Blue1, Patent Blue, Patent Blue VF, Hydrogen [4-[4-(diethylamino)-2',4'-disulphonatobenzhydrylidene]cyclohexa-2,5-dien-1-ylidene]diethylammonium, sodium salt, Patent blue VF sodium salt
18941-0250
18941-1000
TS18941-0250EA
54
USD
TS18941-0250
TS18941-1000
Patent Blue VF (Disulfine blue), pure indicator grade
Patent Blue VF (Disulfine blue)
Formula:
C₂₇H₃₁N₂NaO₆S₂ MW: 566.67 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00012119 CAS Number: 129-17-9 |
Specification Test Results
Appearance | Dark blue-green to dark purple Crystalline powder |
Infrared spectrum | Conforms |
UV spectrum | Complies |
Suitability | (as acid-base indicator) Passes test |
Specific optical rotation | -46° to -52° (20°C, 589 nm) (1% in DMSO) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...