2-Chloro-1-methylpyridinium iodide 97%

Supplier: Thermo Scientific Chemicals
Warning

18926-0050 18926-0250 18926-1000
TS18926-0050EA 29 USD
TS18926-0050 TS18926-0250 TS18926-1000
2-Chloro-1-methylpyridinium iodide 97%
2-Chloro-1-methylpyridinium iodide
Formula: C₆H₇ClIN
MW: 255.49 g/mol
Melting Pt: 207
MDL Number: MFCD00011984
CAS Number: 14338-32-0
EINECS: 238-288-7

Order Now

Specification Test Results

Appearance Yellow Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 195 to 200 °C
Titration Argentometric 96.0 to 104.0 %
Solubility light yellow
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR