Bis(cyclopentadienyl)zirconium dichloride 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
18792-0050
18792-0250
18792-1000
TS18792-0050EA
39
USD
TS18792-0050
TS18792-0250
TS18792-1000
Bis(cyclopentadienyl)zirconium dichloride 98+%
Bis(cyclopentadienyl)zirconium dichloride
Formula:
C₁₀H₁₀Cl₂Zr Melting Pt: 242…245 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00003726 CAS Number: 1291-32-3 EINECS: 215-066-8 |
Specification Test Results
Appearance | White to light beige Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 242 to 245 °C |
Titration Argentometric | ≥98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...