Benzyltributylammonium chloride 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-Benzyl-N,N-dibutylbutan-1-aminium chloride

18615-1000 18615-5000
TS18615-1000EA 40 USD
TS18615-1000 TS18615-5000
Benzyltributylammonium chloride 98+%
Benzyltributylammonium chloride
Formula: C₁₉H₃₄ClN
MW: 311.94 g/mol
Melting Pt: 155…163 °C
MDL Number: MFCD00011849
CAS Number: 23616-79-7
EINECS: 245-787-3

Order Now

Specification Test Results

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Titration Argentometric 98.0 to 102.0 %
Water ≤1 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR