(2-Methoxyethoxy)methyl chloride 94%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Chloromethoxy-2-methoxyethane, 1-Chloro-2,5-dioxahexane, MEM chloride

18598-0100 18598-0250 18598-1000
TS18598-0100EA 79 USD
TS18598-0100 TS18598-0250 TS18598-1000
(2-Methoxyethoxy)methyl chloride 94%
(2-Methoxyethoxy)methyl chloride
Formula: ClCH₂OCH₂CH₂OCH₃
MW: 124.57 g/mol
Boiling Pt: 50…52 °C (13 mmHg)
Density: 1.09 g/cm³
Flash Pt: 54 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000888
CAS Number: 3970-21-6
EINECS: 223-589-8
UN: 1992
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4260 to 1.4280 (20°C, 589 nm)
GC ≥92 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR