Poly(acrylic acid), M.W. ∼ 5,000 50% (w/w) in water

Supplier: Thermo Scientific Chemicals

18500-0010 18500-0050 18500-2500
TS18500-0010EA 270 USD
TS18500-0010 TS18500-0050 TS18500-2500
Poly(acrylic acid), M.W. ∼ 5,000 50% (w/w) in water
Poly(acrylic acid)
Formula: C₃H₄O₂
Boiling Pt: 100 °C (760 mmHg)
Density: 1.18 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00084394
CAS Number: 9003-01-4

Order Now

Specification Test Results

Appearance Clear to hazy liquid
Infrared spectrum Conforms
Assay 48 to 52 % Solids
Molecular weight 3000 to 5000 (approx)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR