Benzotriazole 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,2,3-Benzotriazole, 1H-1,2,3-Benzotriazole, 1H-Benzotriazole, 1H-Benzo[d][1,2,3]triazole

18388-0025 18388-1000 18388-5000
TS18388-0025EA 576 USD
TS18388-0025 TS18388-1000 TS18388-5000
Benzotriazole 99%
Benzotriazole
Formula: C₆H₅N₃
Boiling Pt: 204 °C (15 mmHg)
Melting Pt: 94…99 °C
Flash Pt: 170 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00005699
CAS Number: 95-14-7
EINECS: 202-394-1
UN: 3077
ADR: 9,III
Merck Index: 1301109
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0ba54cc42ef5ad9dbae6d7f118a3af1ae642287&vid=ce57b3f0d2583e32220d1e191d0202ad452e6475&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to yellow to light beige Powder or crystals or granules or needles or flakes
Infrared spectrum Conforms
Melting point 94 to 99 °C
Titration with NaOH ≥98.5 %
Solubility (2 % in water) Clear colorless solution
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR