Pyridinium chlorochromate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: PCC

18367-0250 18367-1000 18367-5000
TS18367-0250EA 36 USD
TS18367-0250 TS18367-1000 TS18367-5000
Pyridinium chlorochromate 98%
Pyridinium chlorochromate
Formula: C₅H₆ClCrNO₃
MW: 215.56 g/mol
Melting Pt: 205 °C
MDL Number: MFCD00013106
CAS Number: 26299-14-9
EINECS: 247-595-5
UN: 1479
ADR: 5.1,II
Merck Index: 1308064

Order Now

Specification Test Results

Appearance Orange Crystalline powder
Infrared spectrum Conforms
Titration Iodometric 97.5 to 102.5 %
HPLC ≥98.5 %
Residue after ignition ≤0.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR