(p-Toluenesulphonyl)methyl isocyanide 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-((Isocyanomethyl)sulphonyl)-4-methylbenzene, Tosylmethyl isocyanide

18245-0050 18245-0250 18245-1000
TS18245-0050EA 46 USD
TS18245-0050 TS18245-0250 TS18245-1000
(p-Toluenesulphonyl)methyl isocyanide 98%
(p-Toluenesulphonyl)methyl isocyanide
Formula: C₉H₉NO₂S
MW: 195.24 g/mol
Melting Pt: 111…113 °C
Storage Temperature: Freezer
MDL Number: MFCD00000005
CAS Number: 36635-61-7
EINECS: 253-140-1
UN: 2811
ADR: 6.1 II

Order Now

Specification Test Results

Appearance Light yellow to beige to light brown Powder and/or chunks
Infrared spectrum Conforms
Melting point 109 to 115 °C (Dec)
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR