Butyl chloroformate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Chloroformic acid butyl ester, n-Butyl chloroformate

18017-0010 18017-0250
TS18017-0010EA 206 USD
TS18017-0010 TS18017-0250
Butyl chloroformate 98%
Butyl chloroformate
Formula: C₅H₉ClO₂
Boiling Pt: 142 °C (1013 hPa)
Melting Pt: –70 °C
Density: 1.06 g/cm³
Flash Pt: 39 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000650
CAS Number: 592-34-7
EINECS: 209-750-5
UN: 2743
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4110 to 1.4130 (20°C, 589 nm)
GC ≥97.5 %
Color scale ≤100 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR