N,N-Diethylaniline borane 97%

Supplier: Thermo Scientific Chemicals
Danger

17677-0250 17677-1000 17677-2500
TS17677-0250EA 73 USD
TS17677-0250 TS17677-1000 TS17677-2500
N,N-Diethylaniline borane 97%
N,N-Diethylaniline borane
Formula: C₁₀H₁₈BN
Melting Pt: 30…27 °C
Density: 0.92 g/cm³
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00013187
CAS Number: 13289-97-9
UN: 3148
ADR: 4.3,I

Order Now

Specification Test Results

Appearance Clear colorless to amber Liquid
Infrared spectrum Conforms
Refractive index 1.5220 to 1.5250 (20°C, 589 nm)
Titration ≥96.0 % (Active hydrogen)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR