Iodosobenzene diacetate 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Iodobenzene diacetate, (Diacetoxyiodo)benzene
17656-0250
17656-1000
17656-5000
TS17656-0250EA
66
USD
TS17656-0250
TS17656-1000
TS17656-5000
Iodosobenzene diacetate 98%
Iodosobenzene diacetate
Formula:
C₁₀H₁₁IO₄ Melting Pt: 161…165 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008692 CAS Number: 3240-34-4 EINECS: 221-808-1 |
Specification Test Results
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 159 to 164 °C |
Titration Iodometric | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...