4-(2-Pyridylazo)resorcinol monosodim salt monohydrate 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
17632-0050
17632-0250
17632-1000
TS17632-0050EA
134
USD
TS17632-0050
TS17632-0250
TS17632-1000
4-(2-Pyridylazo)resorcinol monosodim salt monohydrate 99+%
4-(2-Pyridylazo)resorcinol monosodim salt monohydrate
Formula:
C₁₁H₈N₃NaO₂·1H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00150712 CAS Number: 16593-81-0 EINECS: 236-339-8 |
Specification Test Results
Appearance | Orange-brown to brown Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤10 % (K.F.) |
Solubility | (0.1 % in water) Clear |
Specific extinction E (1%/1cm) | ≥1100 (413 nm, water) |
Titration with HClO4 | ≥99 % (on dry substance) |
Suitability | (for Pb determination) Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...