Nitrosium tetrafluoroborate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Nitrosyl tetrafluoroborate, Nitrosonium tetrafluoroborate

17439-0050 17439-0250 17439-1000
TS17439-0050EA 75 USD
TS17439-0050 TS17439-0250 TS17439-1000
Nitrosium tetrafluoroborate 97%
Nitrosium tetrafluoroborate
Formula: BF₄NO
MW: 116.81 g/mol
Melting Pt: 250 °C (subl.)
Density: 2.185 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00011433
CAS Number: 14635-75-7
EINECS: 238-679-2
UN: 3260
ADR: 8,II

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Assay ≥96.0 % (ex F)
Melting point 75 to 83 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR