Tiron®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,2-Dihydroxybenzene-3,5-disulphonic acid disodium salt, Catechol-3,5-disulphonic acid disodium salt, 4,5-Dihydroxy-1,3-benzenedisulphonic acid disodium salt, 3,5-Pyrocatecholdisulphonic acid disodium salt, Disodium 4,5-dihydroxybenzene-1,3-disulphonate

17414-0250 17414-1000 17414-5000
TS17414-0250EA 51 USD
TS17414-0250 TS17414-1000 TS17414-5000
Tiron®
Tiron
Formula: C₆H₄Na₂O₈S₂
MW: 314.20 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00149531
CAS Number: 149-45-1
EINECS: 205-741-5

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Water 4.5 to 6.0 % (K.F.)
Titration with NaOH ≥98.0 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR