Fluorescein disodium salt, pure
Supplier: Thermo Scientific Chemicals
17324-0025
17324-1000
17324-5000
TS17324-0025EA
426
USD
TS17324-0025
TS17324-1000
TS17324-5000
Fluorescein disodium salt, pure
Fluorescein disodium salt
Formula:
C₂₀H₁₀Na₂O₅ Melting Pt: 320 °C Storage Temperature: Ambient |
MDL Number:
MFCD00167039 CAS Number: 518-47-8 EINECS: 208-253-0 Merck Index: 1304185 |
|
Specification Test Results
Appearance | Orange-red to green-brown to dark brown Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤10 % (105°C) |
UV Lambda max | 488 to 491 nm (in buffer pH 8) |
Absorption Ratio P-15/P+15 | 1.2 to 1.7 |
Specific absorbance A (1%/1cm) | ≥1700 (at lambda max.) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...