Phthalimide potassium salt derivative 99%

Supplier: Thermo Scientific Chemicals

Synonyms: N-Potassium phthalimide, Potassium 1,3-dioxoisoindolin-2-ide, Potassium phthalimide

17086-0025 17086-1000 17086-5000
TS17086-0025EA 331 USD
TS17086-0025 TS17086-1000 TS17086-5000
Phthalimide potassium salt derivative 99%
Phthalimide potassium salt
Formula: C₈H₄KNO₂
MW: 185.22 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00005887
CAS Number: 1074-82-4
EINECS: 214-046-6

Order Now

Specification Test Results

Appearance White to yellow to green Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
Titration with HClO4 ≥98.5 % (On dried substance)
HPLC ≥97.5 %
Specific optical rotation -63° to -67° (20°C, 589 nm) (c=3, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR