1,3-Dibromo-5,5-dimethylhydantoin 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
16954-0025
16954-0250
16954-5000
TS16954-0025EA
271
USD
TS16954-0025
TS16954-0250
TS16954-5000
1,3-Dibromo-5,5-dimethylhydantoin 98%
1,3-Dibromo-5,5-dimethylhydantoin
Formula:
C₅H₆Br₂N₂O₂ Melting Pt: 187…191 °C (dec.) Flash Pt: 155 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003189 CAS Number: 77-48-5 EINECS: 201-030-9 UN: 3085 ADR: 5.1,II |
Specification Test Results
Appearance | Cream to light yellow Powder |
Infrared spectrum | Conforms |
Titration Iodometric | ≥97.5 % |
Loss on drying | ≤0.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...