Ninhydrin, Spectrophotometric Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate
16587-0250
16587-1000
16587-5000
TS16587-0250EA
88
USD
TS16587-0250
TS16587-1000
TS16587-5000
Ninhydrin, Spectrophotometric Grade
Ninhydrin
Formula:
C₉H₆O₄ Melting Pt: 250 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00003791 CAS Number: 485-47-2 EINECS: 207-618-1 Merck Index: 1306582 |
|
Specification Test Results
Appearance | White to light yellow Crystalline powder or crystals |
Infrared spectrum | Conforms |
Sulfated ash | ≤0.1 % |
pH | 4.0 to 5.6 (1 % in water) |
Suitability | (for amino acid analysis) Passes test |
Specific optical rotation | +13.5° to +15.5° (20°C, 589 nm) (c=10, 6 N HCl) (dried substance) |
Titration with HClO4 | 98.5 to 101.0 % (dried substance) |
Sulfates | ≤300 ppm |
Chlorides | ≤200 ppm |
Ammonium (NH4) | ≤200 ppm |
Iron (Fe) | ≤10 ppm |
Ninhydrin-positive substances | ≤0.5 % (Total) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...