EEDQ (ethyl-N-(2-ethoxy-1,2-dihydroquinoline)carboxylate) 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline, N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, Ethyl 2-ethoxyquinoline-1(2H)-carboxylate

16535-0250 16535-1000 16535-2500
TS16535-0250EA 58 USD
TS16535-0250 TS16535-1000 TS16535-2500
EEDQ (ethyl-N-(2-ethoxy-1,2-dihydroquinoline)carboxylate) 99+%
EEDQ (ethyl-N-(2-ethoxy-1,2-dihydroquinoline)carboxylate)
Formula: C₁₄H₁₇NO₃
MW: 247.29 g/mol
Boiling Pt: 125…128 °C (.1 mmHg)
Melting Pt: 63…66 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006703
CAS Number: 16357-59-8
EINECS: 240-418-2

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 63 to 66 °C
HPLC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR