Benzyltriethylammonium bromide 99%

Supplier: Thermo Scientific Chemicals
Warning

16420-0250 16420-1000
TS16420-0250EA 39 USD
TS16420-0250 TS16420-1000
Benzyltriethylammonium bromide 99%
Benzyltriethylammonium bromide
Formula: C₁₃H₂₂BrN
MW: 272.23 g/mol
Melting Pt: 187…197 °C
Storage Temperature: Ambient
MDL Number: MFCD00011822
CAS Number: 5197-95-5
EINECS: 225-986-1

Order Now

Specification Test Results

Appearance White to cream to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 187 to 197 °C (Dec)
Titration Argentometric ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR