Benzyltriethylammonium chloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzyl triethylammonium chloride

16402-0025 16402-1000 16402-5000
TS16402-0025EA 556 USD
TS16402-0025 TS16402-1000 TS16402-5000
Benzyltriethylammonium chloride 98%
Benzyltriethylammonium chloride
Formula: C₁₃H₂₂ClN
MW: 227.78 g/mol
Melting Pt: 185 °C (dec.)
Flash Pt: >275 °C
Storage Temperature: Ambient
MDL Number: MFCD00011824
CAS Number: 56-37-1
EINECS: 200-270-1

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Titration Argentometric 97.5 to 102.5 %
Water ≤2.0 % (K.F.)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR