Chlorosulphonyl isocyanate 98+%

Supplier: Thermo Scientific Chemicals
Danger

16204-0010 16204-0050 16204-0500 16204-2500
TS16204-0010EA 1057 USD
TS16204-0010 TS16204-0050 TS16204-0500 TS16204-2500
Chlorosulphonyl isocyanate 98+%
Chlorosulphonyl isocyanate
Formula: CClNO₃S
MW: 141.53 g/mol
Boiling Pt: 106 °C (760 mmHg)
Melting Pt: –44 °C
Density: 1.626 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011608
CAS Number: 1189-71-5
EINECS: 214-715-2
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4450 to 1.4490 (20°C, 589 nm)
Titration with NaOH ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR