Ponceau S, pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Hydroxy-4-(2-sulpho-4-[4-sulphophenylazo]phenylazo)-2,7-naphthalenedisulphonic acid sodium salt, CI 27195, Ponceau, 3-Hydroxy-4-((2-sulpho-4-((4-sulphophenyl)diazenyl)phenyl)diazenyl)naphthalene-2,7-disulphonic acid, tetrasodium salt
16147-0100
16147-0250
16147-1000
TS16147-0100EA
31
USD
TS16147-0100
TS16147-0250
TS16147-1000
Ponceau S, pure
Ponceau S
Formula:
C₂₂H₁₂N₄Na₄O₁₃S₄ MW: 760.56 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003892 CAS Number: 6226-79-5 EINECS: 228-319-2 |
Specification Test Results
Appearance | Bordeaux to dark brown Crystalline powder |
Infrared spectrum | Conforms |
UV spectrum | Complies |
Dye content | ≥70 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...