N,N-Dimethylformamide dimethyl acetal ca. 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,1-Dimethoxytrimethylamine, Esterate M, N,N-Dimethylformamide Dimethyl Acetal, 1,1-Dimethoxy-N,N-dimethylmethylamine, DMF-DMA

16125-0250 16125-1000 16125-5000
TS16125-0250EA 60 USD
TS16125-0250 TS16125-1000 TS16125-5000
N,N-Dimethylformamide dimethyl acetal ca. 97%
N,N-Dimethylformamide dimethyl acetal
Formula: C₅H₁₃NO₂
MW: 119.16 g/mol
Boiling Pt: 102
Density: 0.89 g/cm³
Flash Pt: 7 °C
MDL Number: MFCD00008482
CAS Number: 4637-24-5
EINECS: 225-063-3
UN: 1993

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3950 to 1.3970 (20°C, 589 nm)
GC ≥96.0 %
Specific optical rotation -53° to -63° (20°C, 589 nm) (c=0.5, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR