N,N,N-Trimethylanilinium tribromide 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Phenyltrimethylammonium tribromide, Mono(N,N,N-trimethylbenzenaminium) tribromide, Trimethylphenylammonium tribromide

16094-0250 16094-1000
TS16094-0250EA 54 USD
TS16094-0250 TS16094-1000
N,N,N-Trimethylanilinium tribromide 97%
N,N,N-Trimethylanilinium tribromide
Formula: C₉H₁₄Br₃N
MW: 375.93 g/mol
Melting Pt: 113…117 °C
MDL Number: MFCD00011789
CAS Number: 4207-56-1
EINECS: 224-127-8
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance Orange Crystalline powder
Infrared spectrum Conforms
Melting point 113 to 117 °C
Titration ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR