Boron trifluoridedimethanol complex (50 - 52% BF₃) (w/w)

Supplier: Thermo Scientific Chemicals
Danger

15890-0010 15890-0500 15890-2500
TS15890-0010EA 104 USD
TS15890-0010 TS15890-0500 TS15890-2500
Boron trifluoridedimethanol complex (50 - 52% BF₃) (w/w)
Boron trifluoridedimethanol complex
Formula: BF₃·2CH₃OH
MW: 131.89 g/mol
Boiling Pt: 59 °C (4 mmHg)
Melting Pt: –20 °C
Density: 1.222 g/cm³
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD00071635
CAS Number: 2802-68-8
UN: 2922
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Assay 50 to 53 wt% (BF3)
Density (20°C) 1.210 to 1.235 kg/l
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤100 ppm
Arsenic (As) ≤1 ppm
Iron (Fe) ≤30 ppm
Sodium (Na) ≤0.1 %
Heavy metals (as Pb) ≤10 ppm
Nitrogen compounds ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR