1,5-Diphenylcarbazide 98% ACS

Supplier: Thermo Scientific Chemicals
Warning

15884-0250 15884-1000 15884-5000
TS15884-0250EA 103 USD
TS15884-0250 TS15884-1000 TS15884-5000
1,5-Diphenylcarbazide 98% ACS
1,5-Diphenylcarbazide
Formula: C₁₃H₁₄N₄O
Melting Pt: 173…176 °C
Storage Temperature: Ambient
MDL Number: MFCD00003013
CAS Number: 140-22-7
EINECS: 205-403-7
Merck Index: 1303353
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03cf8836a36f1077443ec2e3cfa151621925bd46&vid=a1b2bd876c3fbd9d6ded95c8430e4ec16bf5172b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance light grey to light purple to light brown Powder or crystals
Infrared spectrum Conforms
Melting point 173 to 176 °C
Sulfated ash ≤0.05 %
Solubility (in aqueous acetone) Passes test
HPLC ≥97.5 %
Sensitivity (to chromate) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR