Indigo carmine, high purity biological stain, for analysis

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: CI 73015, FD&C Blue #2, 5,5'-Indigodisulphonic acid, Disodium salt, Food blue No. 2, acid Blue 74, Sodium 3,3'-dioxo-[2,2'-biindolinylidene]-5,5'-disulphonate, FD and C Blue No. 2, Carmine Blue

15744-0250 15744-1000 15744-5000
TS15744-0250EA 59 USD
TS15744-0250 TS15744-1000 TS15744-5000
Indigo carmine, high purity biological stain, for analysis
Indigo carmine
Formula: C₁₆H₈N₂Na₂O₈S₂
MW: 466.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005723
CAS Number: 860-22-0
EINECS: 212-728-8
Merck Index: 1304969

Order Now

Specification Test Results

Appearance Dark blue Crystalline powder
Infrared spectrum Conforms
Heavy metals ≤40 ppm
Additional info dinatrium-3,3'-dioxo-2,2'-bisindolylidene-5,7'-disulphonate: 18.0%
Dye content ≥85 %
Arsenic (As) ≤3 ppm
Cadmium (Cd) ≤1 ppm
Lead (Pb) ≤10 ppm
Mercury (Hg) ≤1 ppm
Free amine ≤0.01 % (aromatic)
Insoluble matter ≤0.2 % (in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR