N-Hydroxysuccinimide 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Hydroxypyrrolidine-2,5-dione, NHS, HOSu

15727-0010 15727-0250 15727-1000 15727-2500
TS15727-0010EA 819 USD
TS15727-0010 TS15727-0250 TS15727-1000 TS15727-2500
N-Hydroxysuccinimide 98+%
N-Hydroxysuccinimide
Formula: C₄H₅NO₃
MW: 115.09 g/mol
Melting Pt: 93…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00005516
CAS Number: 6066-82-6
EINECS: 228-001-3

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals (May have a tendency to cake)
Infrared spectrum Conforms
Water ≤2.0 % (K.F.)
Solubility The product may contain some hydroxysuccinimide:ammonia adduct, which is virtually insoluble in acetone
Titration with NaOH 98.0 to 101.0 % (On dried substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR