4-Methylumbelliferone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-MU

15638-0250 15638-1000 15638-5000
TS15638-0250EA 40 USD
TS15638-0250 TS15638-1000 TS15638-5000
4-Methylumbelliferone 97%
4-Methylumbelliferone
Formula: C₁₀H₈O₃
MW: 176.17 g/mol
Melting Pt: 186…191 °C
Storage Temperature: Ambient
MDL Number: MFCD00006866
CAS Number: 90-33-5
EINECS: 201-986-7

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 186 to 191 °C
Water ≤2.5 % (K.F.)
Sulfated ash ≤0.1 %
HPLC ≥96.0 %
Titration with NaOH ≥96.0 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR