Alizarine 97%, pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Alizarin, 1,2-Dihydroxy-9,10-anthraquinone, 1,2-Dihydroxy-9,10-anthracenedione, 1,2-Dihydroxyanthraquinone

15369-0025 15369-0250 15369-1000 15369-5000
TS15369-0025EA 944 USD
TS15369-0025 TS15369-0250 TS15369-1000 TS15369-5000
Alizarine 97%, pure
Alizarine
Formula: C₁₄H₈O₄
MW: 240.22 g/mol
Boiling Pt: 430 °C (1013 hPa)
Melting Pt: 287…289 °C
Storage Temperature: Ambient
MDL Number: MFCD00001201
CAS Number: 72-48-0
EINECS: 200-782-5
Merck Index: 1300246

Order Now

Specification Test Results

Appearance Orange to orange-brown Powder
Infrared spectrum Conforms
Loss on drying ≤2 %
Dye content ≥96.0 %
Residue after evaporation ≤0.002 %
Acidity ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR