Giemsa's stain, pure, high purity biological stain
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Azure mixture sicc. Giemsa stain, Azur-Eosin-Methylene blue acc. Giemsa, Giemsa's azur-eosin-methylene blue
15323-0050
15323-0250
TS15323-0050EA
53
USD
TS15323-0050
TS15323-0250
Giemsa's stain, pure, high purity biological stain
Giemsa's stain
Melting Pt:
300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00081642 CAS Number: 51811-82-6 EINECS: 257-438-2 |
Specification Test Results
Appearance | Dark green to black Crystalline powder or crystals |
Infrared spectrum | Conforms |
Loss on drying | ≤10 % (110°C) |
UV Lambda max | 520 to 525 nm (in MeOH) |
Specific absorbance A (1%/1cm) | ≥600 (in MeOH) On dried substance |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...