Acid green 1, pure

Supplier: Thermo Scientific Chemicals

Synonyms: Naphthol Green B, Iron, tris(5,6-dihydro-5,6-dioxo-2-naphthalenesulphonic acid 5-oximato)-,trisodium salt

15293-0250 15293-1000 15293-5000
TS15293-0250EA 30 USD
TS15293-0250 TS15293-1000 TS15293-5000
Acid green 1, pure
Acid green 1
Formula: C₃₀H₁₈FeN₃O₁₅S₃
MW: 878.45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003886
CAS Number: 19381-50-1

Order Now

Specification Test Results

Appearance Dark green Crystalline powder
Infrared spectrum Conforms
UV spectrum Complies

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR