5-(4-(Dimethylamino)benzylidene)rhodanine 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Dimethylaminobenzalrhodanine, 5-(4-Dimethylaminobenzylidene)-2-thioxo-4-thiazolidinone, p-Dimethylaminobenzalrhodanine, 5-(4-(Dimethylamino)benzylidene)-2-mercaptothiazol-4(5H)-one, p-(Dimethylamino)benzal-5-rhodanine, (5E)-5-[4-(Dimethylamino)benzylidene]-2-mercapto-1,3-thiazol-4(5H)-one

15147-0100 15147-0250 15147-1000
TS15147-0100EA 90 USD
TS15147-0100 TS15147-0250 TS15147-1000
5-(4-(Dimethylamino)benzylidene)rhodanine 99%
5-(4-(Dimethylamino)benzylidene)rhodanine
Formula: C₁₂H₁₂N₂OS₂
Melting Pt: 285…288 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064857
CAS Number: 536-17-4
EINECS: 208-625-2

Order Now

Specification Test Results

Appearance Red Powder
Infrared spectrum Conforms
Solubility (0.03 % in acetone) Clear
Titration with TBAH 98.5 to 101.5 %
Suitability (for Ag determination) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR