Phenol red sodium salt, water soluble, pure indicator

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, sodium salt, Phenol red water soluble, Sodium hydrogen 4,4'-(3H-2,1-benzoxathiol-3-ylidene)bisphenolate S,S-dioxide

15144-0050 15144-0250 15144-1000
TS15144-0050EA 28 USD
TS15144-0050 TS15144-0250 TS15144-1000
Phenol red sodium salt, water soluble, pure indicator
Phenol red sodium salt
Formula: C₁₉H₁₃NaO₅S
MW: 376.35 g/mol
Melting Pt: 285 °C
Storage Temperature: Ambient
MDL Number: MFCD00066901
CAS Number: 34487-61-1
EINECS: 252-057-8
Merck Index: 1207397

Order Now

Specification Test Results

Appearance Dark red to brown Powder
Infrared spectrum Conforms
Solubility (0.1 % in water) Clear
Specific extinction E (1%/1cm) ≥1600 (558 nm, in buffer)
Visual transition interval from pH 6.8 (yellow) to pH 8.2 (red)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR