Phenol red, pure indicator
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
15143-0050
15143-0250
15143-1000
TS15143-0050EA
33
USD
TS15143-0050
TS15143-0250
TS15143-1000
Phenol red, pure indicator
Phenol red
Formula:
C₁₉H₁₄O₅S Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003552 CAS Number: 143-74-8 EINECS: 205-609-7 Merck Index: 1307329 |
Specification Test Results
Appearance | Red Powder |
Infrared spectrum | Conforms |
Solubility | (0.1 % in 95% ethanol) Clear soluble |
UV Lambda max | 555 to 561 nm (in buffer) |
Specific extinction E (1%/1cm) | ≥1700 (at lambda max., in buffer) |
Visual transition interval | from pH 6.8 (yellow) to pH 8.2 (red) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...