Cresol red sodium salt, water soluble, pure indicator

Supplier: Thermo Scientific Chemicals

15139-0050 15139-0250
TS15139-0050EA 30 USD
TS15139-0050 TS15139-0250
Cresol red sodium salt, water soluble, pure indicator
Cresol red sodium salt
Formula: C₂₁H₁₇NaO₅S
Melting Pt: 250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00001618
CAS Number: 62625-29-0

Order Now

Specification Test Results

Appearance Brown Powder
Infrared spectrum Conforms
Solubility (0.1 % in water) Clear
Visual transition interval from pH 7.0 (yellow) to pH 8.8 (purple)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR