Bromophenol blue sodium salt, soluble_water, pure Indicator

Supplier: Thermo Scientific Chemicals

Synonyms: Bromophenol blue sodium, Sodium α-(3,5-dibromo-4-oxo-2,5-cyclohexadienylidene)-α-(3,5-dibromo-4-hydroxyphenyl)toluenesulphonate, Sodium 2,6-dibromo-4-(3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)phenolate, Bromophenol blue water soluble, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2,6-dibromophenol] S,S-dioxide sodium salt

15135-0050 15135-0250 15135-1000
TS15135-0050EA 35 USD
TS15135-0050 TS15135-0250 TS15135-1000
Bromophenol blue sodium salt, soluble_water, pure Indicator
Bromophenol blue sodium salt
Formula: C₁₉H₉Br₄NaO₅S
MW: 691.95 g/mol
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013793
CAS Number: 34725-61-6
EINECS: 252-170-2

Order Now

Specification Test Results

Appearance Dark green to brown-black to blue-black Powder
Infrared spectrum Conforms
UV spectrum Complies
Visual transition interval from pH 3.0 (yellow) to pH 4.6 (blue)
Clarity of solution (0.1% in water) clear


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR