Bromocresol green, pure indicator grade

Supplier: Thermo Scientific Chemicals

Synonyms: 3,3-Bis(3,5-dibromo-4-hydroxy-2-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

15131-0050 15131-0250
TS15131-0050EA 57 USD
TS15131-0050 TS15131-0250
Bromocresol green, pure indicator grade
Bromocresol green
Formula: C₂₁H₁₄Br₄O₅S
Melting Pt: 225 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005874
CAS Number: 76-60-8
EINECS: 200-972-8
Merck Index: 1301370
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12b4d4b3d992f8e86746537bf756af442752c853&vid=553ef1836a5b191a43558f8550e78cf2e424446e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Yellow to beige to light brown Powder
Infrared spectrum Conforms
Solubility (0.1 % in ethanol) Clear
UV Lambda max 613 to 621 nm (in buffer)
Specific extinction E (1%/1cm) ≥550 (at lambda max)
Visual transition interval from pH 3.8 (yellow) to pH 5.4 (blue)
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR