Chromotrope 2B, pure

Supplier: Thermo Scientific Chemicals
Warning

14975-0050 14975-0250
TS14975-0050EA 24 USD
TS14975-0050 TS14975-0250
Chromotrope 2B, pure
Chromotrope 2B
Formula: C₁₆H₉N₃Na₂O₁₀S₂
MW: 513.37 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00003951
CAS Number: 548-80-1

Order Now

Specification Test Results

Appearance Dark green to greenish-brown to brown Powder
Infrared spectrum Conforms
Loss on drying ≤18 %
UV Lambda max 513 to 517 nm (c=0.1, H2O)
Specific extinction E (1%/1cm) ≥580 (on dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR