Trimethyl orthoformate 99%

Supplier: Thermo Scientific Chemicals
Danger

14866-0010 14866-0050 14866-1000 14866-5000
TS14866-0010EA 84 USD
TS14866-0010 TS14866-0050 TS14866-1000 TS14866-5000
Trimethyl orthoformate 99%
Trimethyl orthoformate
Formula: C₄H₁₀O₃
MW: 106.12 g/mol
Boiling Pt: 101…102 °C (760 mmHg)
Melting Pt: –53 °C
Density: 0.97 g/cm³
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00008483
CAS Number: 149-73-5
EINECS: 205-745-7
UN: 3271
ADR: 3,II
Merck Index: 1207012

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3780 to 1.3800 (20°C, 589 nm)
GC ≥98.5 %
Elemental Analysis 8.51 to 9.21 % (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR